Methyl 2-methyl-2-(4-nitro-1H-imidazol-1-yl)propanoate ≥95%
Supplier: Ambeed
|
Warning
|
A835510-1G
A835510-25G
A835510-5G
77622-998EA
8.24
USD
77622-998
77623-000
77623-002
Methyl 2-methyl-2-(4-nitro-1H-imidazol-1-yl)propanoate ≥95%
Methyl 2-methyl-2-(4-nitro-1H-imidazol-1-yl)propanoate
Precaución: for Research Use Only
Formula:
C₈H₁₁N₃O₄ MW: 213.1906 g/mol |
MDL Number:
MFCD28385911 CAS Number: 865774-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...