2-Bromo-5-(bromomethyl)pyridine ≥98%

Supplier: Ambeed
Danger

A835466-1G A835466-100MG
77595-328EA 110.7 USD
77595-328 77674-238
2-Bromo-5-(bromomethyl)pyridine ≥98%
2-Bromo-5-(bromomethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD11656267
CAS Number: 101990-45-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR