2-(6-Chloropyrazin-2-yl)acetic acid ≥97%
Supplier: Ambeed
|
Warning
|
A835238-1G
A835238-250MG
77670-204EA
146
USD
77670-204
77873-303
2-(6-Chloropyrazin-2-yl)acetic acid ≥97%
2-(6-Chloropyrazin-2-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₂ |
MDL Number:
MFCD09907855 CAS Number: 930798-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...