(S)-2-(((tert-Butyldiphenylsilyl)oxy)methyl)pyrrolidine ≥95%

Supplier: Ambeed
Warning

A834382-1G A834382-250MG A834382-100MG
77599-146EA 105.58 USD
77599-146 77599-148 77622-996
(S)-2-(((tert-Butyldiphenylsilyl)oxy)methyl)pyrrolidine ≥95%
(S)-2-(((tert-Butyldiphenylsilyl)oxy)methyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₂₁H₂₉NOSi
MW: 339.5466 g/mol
MDL Number: MFCD17018324
CAS Number: 209627-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR