4-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)butanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-Me-Abu(4)-OH

A834122-1G A834122-250MG A834122-5G
77686-861EA 33 USD
77686-861 77901-101 77900-598
4-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)butanoic acid ≥98%
4-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₄
MW: 339.38 g/mol
MDL Number: MFCD08275875
CAS Number: 221124-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR