2-(4-Methylpiperazin-1-yl)pyridine-4-boronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine, 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine

A833902-100MG A833902-250MG
77893-626EA 62 USD
77893-626 77893-627
2-(4-Methylpiperazin-1-yl)pyridine-4-boronic acid, pinacol ester ≥98%
2-(4-Methylpiperazin-1-yl)pyridine-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₆BN₃O₂
MDL Number: MFCD05864305
CAS Number: 832114-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR