trans-2-(4-Biphenyl)vinylboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (E)-(2-([1,1'-Biphenyl]-4-yl)vinyl)boronic acid

A833158-250MG A833158-5G A833158-1G
77614-494EA 30.9 USD
77614-494 77684-794 77684-793
trans-2-(4-Biphenyl)vinylboronic acid ≥95%
trans-2-(4-Biphenyl)vinylboronic acid

Precaución: For research use only.

Formula: C₁₄H₁₃BO₂
MW: 224.600 g/mol
MDL Number: MFCD04974059
CAS Number: 352530-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR