(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-(5,5'-Dichloro-6,6'-dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphine), (R)-Cl-MeO-BIPHEP

A832936-100MG A832936-250MG
77595-326EA 144.53 USD
77595-326 77614-492
(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥98%
(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₈H₃₀Cl₂O₂P₂
MDL Number: MFCD03426987
CAS Number: 185913-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR