DL-Asparagine ≥99%

Supplier: Ambeed
Warning

Synonyms: 2,4-Diamino-4-oxobutanoic acid, H-DL-Asn-OH, DL-Aspartic acid 4-amide, (±)-2-Aminosuccinic acid 4-amide, DL-2-Aminosuccinamic acid

A832664-100G A832664-25G A832664-500G A832664-5G
77686-856EA 31 USD
77686-856 77686-857 77686-858 77901-317
DL-Asparagine ≥99%
DL-Asparagine

Precaución: for Research Use Only

Formula: C₄H₈N₂O₃
MW: 132.12 g/mol
MDL Number: MFCD00151039
CAS Number: 3130-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR