3-Iodo-2-methylpyridine ≥95%
Supplier: Ambeed
|
Warning
|
A832471-5G
A832471-1G
A832471-100MG
A832471-250MG
77670-194EA
382.81
USD
77670-194
77670-192
77873-298
77873-299
3-Iodo-2-methylpyridine ≥95%
3-Iodo-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₆IN |
MDL Number:
MFCD09702485 CAS Number: 15112-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...