4-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine ≥97%

Supplier: Ambeed
Warning

A832165-100MG A832165-250MG
77749-624EA 73 USD
77749-624 77749-625
4-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine ≥97%
4-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD25949584
CAS Number: 1289150-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR