3,5-Diiodoaniline ≥97%

Supplier: Ambeed
Warning

A830824-1G A830824-5G
77893-621EA 226 USD
77893-621 77893-622
3,5-Diiodoaniline ≥97%
3,5-Diiodoaniline

Precaución: for Research Use Only

Formula: C₆H₅I₂N
MDL Number: MFCD01851092
CAS Number: 35122-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR