N-Ethylglycine ≥98%

Supplier: Ambeed
Warning

A830306-250MG A830306-25G A830306-5G A830306-1G A830306-100MG
77603-112EA 27.68 USD
77603-112 77603-114 77603-116 77670-190 77880-631
N-Ethylglycine ≥98%
N-Ethylglycine

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD00037794
CAS Number: 627-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR