N-((1R,2R)-2-Aminocyclohexyl)-4-methylbenzenesulfonamide ≥97%

Supplier: Ambeed
Warning

A829971-5G A829971-1G
77747-587EA 51 USD
77747-587 77747-586
N-((1R,2R)-2-Aminocyclohexyl)-4-methylbenzenesulfonamide ≥97%
N-((1R,2R)-2-Aminocyclohexyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂S
MW: 268.37 g/mol
MDL Number: MFCD09842716
CAS Number: 174291-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR