2,4-Difluoro-6-iodoaniline ≥95%
Supplier: Ambeed
|
Warning
|
A829774-1G
A829774-250MG
A829774-5G
77614-486EA
29.87
USD
77614-486
77614-488
77886-823
2,4-Difluoro-6-iodoaniline ≥95%
2,4-Difluoro-6-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₄F₂IN |
MDL Number:
MFCD08059536 CAS Number: 582319-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...