5-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A829730-250MG A829730-5G
77684-787EA 55 USD
77684-787 77684-788
5-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥98%
5-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Precaución: For research use only.

Formula: C₇H₄ClN₃O₂
MW: 197.579 g/mol
MDL Number: MFCD20482240
CAS Number: 1315364-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR