β-L-Cytidine ≥95%

Supplier: Ambeed
Warning

A829437-100MG A829437-250MG A829437-5G
77627-624EA 42.23 USD
77627-624 77627-626 77627-628
β-L-Cytidine ≥95%
β-L-Cytidine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₅
MDL Number: MFCD02683605
CAS Number: 26524-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR