Uridine-5'-diphosphoglucuronic acid trisodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium (2S,3S,4S,5R,6R)-6-(((((((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate, UDP-glucuronic acid trisodium, UDP-GlcA, Tripotassium 6-[({[5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphinato)oxy]-3,4,5-trihydroxyoxane-2-carboxylate, UDP-D-GLUCURONIDE TRISODIUM SALT, UDPGA

A828611-100MG A828611-1G A828611-250MG A828611-50MG A828611-5G
77595-318EA 342.35 USD
77595-318 77603-106 77670-182 77670-184 77670-186
Uridine-5'-diphosphoglucuronic acid trisodium salt ≥98%
Uridine-5'-diphosphoglucuronic acid trisodium salt

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₂Na₃O₁₈P₂
MW: 646.23 g/mol
MDL Number: MFCD00037687
CAS Number: 63700-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR