1-Bromo-3,5-dichloro-2-fluorobenzene ≥98%

Supplier: Ambeed
Warning

A828369-1G A828369-250MG
77901-181EA 28 USD
77901-181 77901-003
1-Bromo-3,5-dichloro-2-fluorobenzene ≥98%
1-Bromo-3,5-dichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MDL Number: MFCD11845990
CAS Number: 1160573-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR