5-Fluoro-2-methylbenzothiazole ≥98%

Supplier: Ambeed
Warning

A827653-1G A827653-5G
77878-678EA 44 USD
77878-678 77878-679
5-Fluoro-2-methylbenzothiazole ≥98%
5-Fluoro-2-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₆FNS
MDL Number: MFCD00135018
CAS Number: 399-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR