5-Bromo-2,3-dihydro-1H-inden-1-amine ≥95%

Supplier: Ambeed
Warning

A826117-1G A826117-250MG
77747-579EA 75 USD
77747-579 77747-580
5-Bromo-2,3-dihydro-1H-inden-1-amine ≥95%
5-Bromo-2,3-dihydro-1H-inden-1-amine

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
MW: 212.09 g/mol
MDL Number: MFCD07373956
CAS Number: 185122-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR