2-Pyrid-3-ylbenzoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-3-yl)benzoic acid

A825596-1G A825596-5G
77573-634EA 37.08 USD
77573-634 77603-104
2-Pyrid-3-ylbenzoic acid ≥98%
2-Pyrid-3-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD03426511
CAS Number: 134363-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR