2,4,5-Trimethyl-1H-imidazole ≥95%

Supplier: Ambeed
Danger

A825397-100MG A825397-1G
77749-614EA 75 USD
77749-614 77749-615
2,4,5-Trimethyl-1H-imidazole ≥95%
2,4,5-Trimethyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.1570 g/mol
MDL Number: MFCD00128183
CAS Number: 822-90-2
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR