5-Bromo-4-cyclopropyl-6-methoxypyrimidine ≥95%

Supplier: Ambeed
Warning

A825264-100MG A825264-1G
77599-136EA 20.5 USD
77599-136 77784-155
5-Bromo-4-cyclopropyl-6-methoxypyrimidine ≥95%
5-Bromo-4-cyclopropyl-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
MDL Number: MFCD31556944
CAS Number: 1649454-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR