5-Bromo-4-chloro-6-cyclopropylpyrimidine ≥95%

Supplier: Ambeed
Warning

A825263-5G A825263-1G
77573-632EA 346.08 USD
77573-632 77622-980
5-Bromo-4-chloro-6-cyclopropylpyrimidine ≥95%
5-Bromo-4-chloro-6-cyclopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₆BrClN₂
MW: 233.49 g/mol
MDL Number: MFCD11047047
CAS Number: 27192-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR