1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥98%

Supplier: Ambeed
Warning

A823425-1G A823425-100MG A823425-250MG
77749-611EA 325 USD
77749-611 77749-610 77749-612
1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥98%
1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₅H₂₁BN₂O₂
MW: 272.15 g/mol
MDL Number: MFCD31537145
CAS Number: 2304008-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR