(E)-4-((tert-Butoxycarbonyl)(methyl)amino)but-2-enoic acid ≥95%
Supplier: Ambeed
|
Warning
|
A822816-100MG
A822816-1G
A822816-250MG
77614-472EA
221.45
USD
77614-472
77686-849
77686-850
(E)-4-((tert-Butoxycarbonyl)(methyl)amino)but-2-enoic acid ≥95%
(E)-4-((tert-Butoxycarbonyl)(methyl)amino)but-2-enoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ MW: 215.24 g/mol |
CAS Number:
915788-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...