(1R,5S,6r)-tert-Butyl 6-formyl-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥97%

Supplier: Ambeed
Warning

A821772-5G A821772-1G
77573-614EA 154.5 USD
77573-614 77684-776
(1R,5S,6r)-tert-Butyl 6-formyl-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥97%
(1R,5S,6r)-tert-Butyl 6-formyl-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₇NO₃
MW: 211.25 g/mol
MDL Number: MFCD18782860
CAS Number: 419572-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR