Diethyl sulphone ≥98%

Supplier: Ambeed
Warning

Synonyms: Diethylsulphone, Ethyl sulphone

A821625-1G A821625-250MG A821625-5G
77627-622EA 30.9 USD
77627-622 77880-623 77880-624
Diethyl sulphone ≥98%
Diethyl sulphone

Precaución: for Research Use Only

Formula: C₄H₁₀O₂S
MDL Number: MFCD00007569
CAS Number: 597-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR