tert-Butyl 6-((methylsulfonyl)oxy)-2-azaspiro[3.3]heptane-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A821464-100MG A821464-1G A821464-250MG A821464-5G
77599-126EA 95.33 USD
77599-126 77599-128 77599-130 77784-147
tert-Butyl 6-((methylsulfonyl)oxy)-2-azaspiro[3.3]heptane-2-carboxylate ≥95%
tert-Butyl 6-((methylsulfonyl)oxy)-2-azaspiro[3.3]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅S
MDL Number: MFCD29085851
CAS Number: 1239320-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR