Cefodizime sodium ≥97%

Supplier: Ambeed
Warning

Synonyms: Sodium (6R,7R)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((5-(carboxylatomethyl)-4-methylthiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A820440-10MG A820440-1MG
77595-308EA 8.2 USD
77595-308 77627-618
Cefodizime sodium ≥97%
Cefodizime sodium

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₆Na₂O₇S₄
MW: 628.63 g/mol
MDL Number: MFCD00897059
CAS Number: 86329-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR