N-Fmoc-N-benzyl-glycine ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(benzyl)amino)acetic acid, Fmoc-N-Bzl-Gly-OH

A820102-1G A820102-5G A820102-100G A820102-25G
77573-606EA 17 USD
77573-606 77573-608 77886-811 77886-812
N-Fmoc-N-benzyl-glycine ≥97%
N-Fmoc-N-benzyl-glycine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.428 g/mol
MDL Number: MFCD04112686
CAS Number: 141743-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR