Hexachloroiridic(IV) acid hydrate ≥98%

Supplier: Ambeed
Danger

Synonyms: Dihydrogen hexachloroiridate(IV) xhydrate (Chunks or pellets), Hydrogen iridium hexachloride, Hydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Dihydrogen hexachloroiridate(IV) hydrate

A819984-100MG A819984-250MG A819984-5G A819984-1G
77599-116EA 44.08 USD
77599-116 77618-302 77618-304 77618-300
Hexachloroiridic(IV) acid hydrate ≥98%
Hexachloroiridic(IV) acid hydrate

Precaución: for Research Use Only

Formula: H₂Cl₆Ir·xH₂O
MW: 406.93 g/mol (anhy)
MDL Number: MFCD00149906
CAS Number: 110802-84-1
UN: 3260
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR