Tributyl acetylcitrate ≥97%

Supplier: Ambeed
Warning

Synonyms: Tributyl O-Acetylcitrate, Acetyltributylcitrate, Acetyl tributyl citrate, Tributyl 2-acetoxypropane-1,2,3-tricarboxylate, O-Acetylcitric acid tributyl ester

A819707-1000G A819707-100G A819707-25G A819707-500G
77603-090EA 17.43 USD
77603-090 77603-092 77603-094 77603-096
Tributyl acetylcitrate ≥97%
Tributyl acetylcitrate

Precaución: for Research Use Only

Formula: C₂₀H₃₄O₈
MW: 402.49 g/mol
MDL Number: MFCD00043554
CAS Number: 77-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR