Cefsulodin sodium salt ≥98%

Supplier: Ambeed
Warning

A818560-1MG A818560-100MG A818560-1G
77614-470EA 5.15 USD
77614-470 77901-398 77901-422
Cefsulodin sodium salt ≥98%
Cefsulodin sodium salt

Precaución: for Research Use Only

Formula: C₂₂H₁₉N₄NaO₈S₂
MDL Number: MFCD07793338
CAS Number: 52152-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR