LX-2343 99+%

Supplier: Ambeed
Warning

Synonyms: LX2343, N-(benzo[d][1,3]dioxol-5-yl)-2-(N-(5-chloro-2-methoxyphenyl)phenylsulphonamido)acetamide

A817166-10MG A817166-1MG A817166-50MG A817166-5MG A817166-100MG
77618-284EA 49.44 USD
77618-284 77618-286 77618-288 77618-290 77749-606
LX-2343 99+%
LX-2343

Precaución: for Research Use Only

Formula: C₂₂H₁₉ClN₂O₆S
MW: 474.91 g/mol
MDL Number: MFCD02578482
CAS Number: 333745-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR