5-Amino-2-(trifluoromethoxy)benzotrifluoride ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethoxy)-3-(trifluoromethyl)aniline

A816827-100MG A816827-250MG A816827-1G
77595-304EA 71.75 USD
77595-304 77595-306 77599-112
5-Amino-2-(trifluoromethoxy)benzotrifluoride ≥98%
5-Amino-2-(trifluoromethoxy)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₅F₆NO
MW: 245.12 g/mol
MDL Number: MFCD04972664
CAS Number: 104678-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR