Quinoxalin-6-ylboronic acid ≥98%

Supplier: Ambeed
Warning

A816772-100MG A816772-1G A816772-250MG
77873-281EA 74 USD
77873-281 77873-282 77873-283
Quinoxalin-6-ylboronic acid ≥98%
Quinoxalin-6-ylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BN₂O₂
MDL Number: MFCD08277243
CAS Number: 852432-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR