2-(Chloro(phenyl)methylene)malononitrile 97

Supplier: Ambeed
Danger

A816663-100MG A816663-1G A816663-250MG
77878-665EA 142 USD
77878-665 77878-666 77878-667
2-(Chloro(phenyl)methylene)malononitrile 97
2-(Chloro(phenyl)methylene)malononitrile

Precaución: for Research Use Only

Formula: C₁₀H₅ClN₂
MW: 188.61 g/mol
MDL Number: MFCD00134407
CAS Number: 18270-61-6
UN: 2923
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR