(1-(tert-Butoxycarbonyl)-1H-indol-5-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A816055-1G A816055-250MG A816055-100MG
77886-804EA 80 USD
77886-804 77886-805 77886-803
(1-(tert-Butoxycarbonyl)-1H-indol-5-yl)boronic acid ≥95%
(1-(tert-Butoxycarbonyl)-1H-indol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₄
MDL Number: MFCD06738850
CAS Number: 317830-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR