6-Chloro-3-(cyclopentylmethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide ≥98%

Supplier: Ambeed
Warning

A815426-10G A815426-1G A815426-5G A815426-250MG
77603-064EA 435.63 USD
77603-064 77603-066 77603-068 77880-616
6-Chloro-3-(cyclopentylmethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide ≥98%
6-Chloro-3-(cyclopentylmethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulfonamide 1,1-dioxide

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClN₃O₄S₂
MDL Number: MFCD00865825
CAS Number: 742-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR