6-Chloro-3-fluoropyridin-2-amine ≥97%

Supplier: Ambeed
Warning

A814286-1G A814286-250MG
77618-280EA 227.63 USD
77618-280 77747-569
6-Chloro-3-fluoropyridin-2-amine ≥97%
6-Chloro-3-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂
MDL Number: MFCD18250471
CAS Number: 1260672-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR