1-Bromo-5-fluoropentane ≥98%

Supplier: Ambeed
Danger

A813976-10G A813976-25G A813976-100G
77674-200EA 6 USD
77674-200 77674-202 77882-735
1-Bromo-5-fluoropentane ≥98%
1-Bromo-5-fluoropentane

Precaución: for Research Use Only

Formula: C₅H₁₀BrF
MDL Number: MFCD01709395
CAS Number: 407-97-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR