3-Aminobicyclo[1.1.1]pentane-1-carbonitrile ≥98%

Supplier: Ambeed
Danger

A813227-1G A813227-5G
77622-972EA 580.92 USD
77622-972 77784-126
3-Aminobicyclo[1.1.1]pentane-1-carbonitrile ≥98%
3-Aminobicyclo[1.1.1]pentane-1-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₈N₂
MDL Number: MFCD30719357
CAS Number: 1936643-30-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR