7-Chloro-6-fluoro-3,4-dihydronaphthalen-1(2H)-one ≥95%

Supplier: Ambeed
Warning

A810882-1G A810882-250MG A810882-100MG
77618-272EA 563.41 USD
77618-272 77749-599 77749-598
7-Chloro-6-fluoro-3,4-dihydronaphthalen-1(2H)-one ≥95%
7-Chloro-6-fluoro-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈ClFO
MW: 198.6200 g/mol
MDL Number: MFCD18208404
CAS Number: 1260010-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR