3-((5-Fluoro-4-(4-methyl-2-(methylamino)thiazol-5-yl)pyrimidin-2-yl)amino)benzenesulfonamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Asnuciclib

A810459-1MG A810459-2MG A810459-5MG A810459-10MG
77618-266EA 33.99 USD
77618-266 77747-558 77747-559 77747-557
3-((5-Fluoro-4-(4-methyl-2-(methylamino)thiazol-5-yl)pyrimidin-2-yl)amino)benzenesulfonamide ≥98%
3-((5-Fluoro-4-(4-methyl-2-(methylamino)thiazol-5-yl)pyrimidin-2-yl)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₅H₁₅FN₆O₂S₂
MW: 394.45 g/mol
MDL Number: MFCD28386217
CAS Number: 1421693-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR