5-Amino-1,3-dihydro-2,1-benzoxaborol-1-ol ≥95%

Supplier: Ambeed
Warning

A809629-250MG A809629-5G
77573-586EA 84 USD
77573-586 77784-111
5-Amino-1,3-dihydro-2,1-benzoxaborol-1-ol ≥95%
5-Amino-1,3-dihydro-2,1-benzoxaborol-1-ol

Precaución: for Research Use Only

Formula: C₇H₈BNO₂
MDL Number: MFCD22572914
CAS Number: 947165-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR