1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole ≥98%

Supplier: Ambeed
Warning

A808648-100MG A808648-250MG A808648-1G
77618-260EA 142.14 USD
77618-260 77618-262 77749-593
1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole ≥98%
1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₃H₁₈BN₃O₂
MW: 259.11 g/mol
MDL Number: MFCD22628498
CAS Number: 1362243-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR