3-Methyl-4-(trifluoromethyl)aniline ≥98%

Supplier: Ambeed
Warning

A808493-1G A808493-5G
77684-760EA 139 USD
77684-760 77684-761
3-Methyl-4-(trifluoromethyl)aniline ≥98%
3-Methyl-4-(trifluoromethyl)aniline

Precaución: For research use only.

Formula: C₈H₈F₃N
MW: 175.15 g/mol
MDL Number: MFCD18399852
CAS Number: 106877-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR