Azomethine H sodium salt hydrate ≥97%

Supplier: Ambeed
Warning

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

A808324-1G A808324-5G
77614-438EA 16.48 USD
77614-438 77684-759
Azomethine H sodium salt hydrate ≥97%
Azomethine H sodium salt hydrate

Precaución: For research use only.

Formula: C₁₇H₁₂NNaO₈S₂·xH₂O
MW: 463.42 g/mol
MDL Number: MFCD00149588
CAS Number: 206752-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR